Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAKNRRDRNS WGGFSEKTYE WSSEEEEPVK KAGPVQVLIV KDDHSFELDE TALNRILLSE AVRDKEVVAV SVAGAFRKGK SFLMDFMLRY MYNQESVDWV
The protein encoded by this gene is a GTPase and a Golgi body transmembrane protein. The encoded protein can form a homotetramer and has been shown to interact with spastin and with mitogen-activated protein kinase kinase kinase kinase 4. This protein may be involved in axonal maintenance as evidenced by the fact that defects in this gene are a cause of spastic paraplegia type 3. Three transcript variants encoding two different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATL1 alpha; ATL1 beta; ATL1 gamma; ATL1-delta; Atlastin-1; Bcl11B; BCL11B/TRDC fusion; Brain-specific GTP-binding protein; Ctip 2; CTIP-2; CTIP2; GBP-3; GTP-binding protein 3; Guanine nucleotide-binding protein 3; guanylate-binding protein 3; hGBP3; hRIT1 alpha; Rit 1; Rit1; Spastic paraplegia 3 protein A; Spastic paraplegia 3A homolog; ZNF856B
Gene Aliases: AD-FSP; ATL1; atlastin1; FSP1; GBP3; HSN1D; SPG3; SPG3A
UniProt ID: (Human) Q8WXF7, (Rat) Q6PST4
Entrez Gene ID: (Human) 51062, (Rat) 362750
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support