Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: SEKPSTAHHR PLRSIARFSS KSKSMDKSDE ELQFPKELME DWSTMEVCVD CKKFISEIIS SSRRSLVLAN KRARLKRKTQ SFYMSSPGPS EYCPSERTIS EI
Acts as a actin nucleation factor, remains associated with the slow-growing pointed end of the new filament. Involved in intracellular vesicle transport along actin fibers, providing a novel link between actin cytoskeleton dynamics and intracellular transport. Required for asymmetric spindle positioning and asymmetric cell division during meiosis. Required for normal formation of the cleavage furrow and for polar body extrusion during female germ cell meiosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein spire homolog 1; Spir-1; spire actin nucleation factor 1; spire family actin nucleation factor 1; spire homolog 1; spire-type actin nucleation factor 1; SPIRE1
Gene Aliases: KIAA1135; Spir-1; SPIR1; SPIRE1
UniProt ID: (Human) Q08AE8
Entrez Gene ID: (Human) 56907
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support