Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Cell adhesion protein that promotes adhesion and outgrowth of hippocampal embryonic neurons. Binds directly to bacteria and their components and functions as an opsonin for macrophage phagocytosis of bacteria. Essential in the initiation of the innate immune response and represents a unique pattern-recognition molecule in the ECM for microbial pathogens. Binds bacterial lipopolysaccharide (LPS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Differentially expressed in cancerous and non-cancerous lung cells 1; DIL-1; DKFZp686G21139; F-spondin; FLJ16313; FLJ22401; FLJ34460; Mindin; spondin 2, extracellular matrix protein; Spondin-2
Gene Aliases: 2310045I24Rik; AI504350; DIL-1; DIL1; M-SPONDIN; MINDIN; Mspondin; SPON2; UNQ435/PRO866
UniProt ID: (Human) Q9BUD6, (Mouse) Q8BMS2, (Rat) Q9WV75
Entrez Gene ID: (Human) 10417, (Mouse) 100689, (Rat) 171569
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support