Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: K562 cell lysate, LOVO cell lysate.
Predicted band size: 42 kDa
Subcellular Location: Nucleus, Nucleus speckle.
Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination of target proteins, leading most often to their proteasomal degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HIB homolog 1; PDX-1 C-terminal-interacting factor 1; Roadkill homolog 1; speckle type POZ protein; Speckle-type POZ protein
Gene Aliases: AI315626; BTBD32; Pcif1; SPOP; TEF2
UniProt ID: (Human) O43791, (Mouse) Q6ZWS8
Entrez Gene ID: (Human) 8405, (Mouse) 20747, (Rat) 287643
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support