Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: HASGNGTTKD YCMLYNPYWT ALPSTLENAT SISLMNLTST PLCNLSDIPP VGIKSKAVVV PWGSCHFLEK ARIAQKGGAE AMLVVNNSVL FPPSGNRSE
By searching sequence databases for homologs of Dictyostelium discoideum Impas, which shares homology with presenilin, followed by PCR of lymphocyte and hippocampus cDNA libraries, Grigorenko et al. (2002) cloned SPPL2a. This protein, which may act as an intramembrane protease, contains an N-terminal protease-associated (PA) domain, several transmembrane regions, a hydrophilic loop, conservative sequences around the first and second aspartate residues, and an invariant PAL motif near the C terminus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IMP-3; intramembrane cleaving protease; Intramembrane protease 3; Presenilin-like protein 2; Signal peptide peptidase-like 2A; SPP-like 2A; SPPL2a protein
Gene Aliases: IMP3; PSEC0147; PSL2; SPPL2A
UniProt ID: (Human) Q8TCT8
Entrez Gene ID: (Human) 84888
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support