Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Serine palmitoyltransferase (SPT) is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hLCB1; HSAN; LCB 1; Long chain base biosynthesis protein 1; MGC14645; RP11-118F2.1; serine C-palmitoyltransferase; Serine palmitoyltransferase 1; serine palmitoyltransferase subunit 1; serine palmitoyltransferase, long chain base subunit 1; Serine-palmitoyl-CoA transferase 1; SPT 1; SPT1
Gene Aliases: AW552086; C77762; E030036H05; HSAN1; HSN1; LBC1; LCB1; rCG_44191; RGD1306617; SPT1; SPTI; SPTLC1
UniProt ID: (Human) O15269, (Mouse) O35704, (Rat) D4A2H2
Entrez Gene ID: (Human) 10558, (Mouse) 268656, (Rat) 361213
Molecular Function:
transaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support