Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWTFLGIATF TYFYKKFGDF ITLANREVLL CVLVFLSLGL VLSYRCRHRN GGLLGRQQSG SQFALFSDIL SGLPFIGFFW AKSPPESENK EQLEARRRRK GTNISETSLI GTAACTSTSS QNDPEVIIVG AGVLGSALAA VLSRDGRKVT VIERDLKEPD RIVGEFLQPG GYHVLKDLGL GDTVEGLDAQ VVNGYMIHDQ ESKSEVQIPY PLSENNQVQS GRAFHHGRFI MSLRKAAMAE PNAKFIEGVV LQLLEEDDVV MGVQYKDKET GDIKELHAPL TVVADGLFSK FRKSLVSNKV SVSSHFVGFL MKNAPQFKAN HAELILANPS PVLIYQISSS ETRVLVDIRG EMPRNLREYM VEKIYPQIPD HLKEPFLEAT DNSHLRSMPA SFLPPSSVKK RGVLLLGDAY NMRHPLTGGG MTVAFKDIKL WRKLLKGIPD LYDDAAIFEA KKSFYWARKT SHSFVVNILA QALYELFSAT DDSLHQLRKA CFLYFKLGGE CVAGPVGLLS VLSPNPLVLI GHFFAVAIYA VYFCFKSEPW ITKPRALLSS GAVLYKACSV IFPLIYSEMK YMVH
SQLE, also named as ERG1, SE and SM, belongs to the squalene monooxygenase family. It catalyzes the first oxygenation step in cholesterol synthesis, acting on squalene before cyclization into the basic steroid structure. SQLE may serve as a flux-controlling enzyme beyond 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGR, considered as rate limiting). It is also post translationally regulated by cholesterol-dependent proteasomal degradation. The MW of SQLE is about 64 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ30795; SE; Squalene epoxidase; Squalene monooxygenase
Gene Aliases: ERG1; SQLE
UniProt ID: (Human) Q14534
Entrez Gene ID: (Human) 6713
Molecular Function:
oxidoreductase
oxygenase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support