Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Sterol regulatory element-binding proteins (SREBPs) are transcription factors that are members of the basic helix-loop-helix leucine zipper family of DNA binding proteins. Three isoforms have been identified in mammalian tissues that vary in structure, regulation, and function. SREBP-1a and SREBP-1c (originally cloned as ADD1) are protein products of alternative promoter usage of the SREBP-1 gene. The third isoform is transcribed from a different gene, SREBP-2. SREBPs are present as 120 kDa inactive precursors in the endoplasmic reticulum (ER) membrane. Upon activation, the SREBP protein is translocated to the Golgi and proteolytic cleavage occurs resulting in a mature transcriptionally active 60-78 kDa fragment. In liver and adipose tissues, SREBPs have a significant influence on lipid and cholesterol accumulation by inducing the transcription of genes involved in these processes. While SREBP-1 is thought to be more important in regulating the expression of genes involved in triglyceride synthesis and accumulation, SREBP-2 has been more closely linked to those involved in cholesterol synthesis and accumulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHd1; Class D basic helix-loop-helix protein 1; nt-SREBP1; SREBP-1; Sterol regulatory element-binding protein 1; Sterol regulatory element-binding transcription factor 1
Gene Aliases: BHLHD1; SREBF1; SREBP-1c; SREBP1; SREBP1a
UniProt ID: (Human) P36956
Entrez Gene ID: (Human) 6720
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support