Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: HMMYPSPHAV MYAPTSGLGD GSLTVLNAFS QAPSTMQVSH SQVQEPGGVP QVFLTASSGT VQIPVSAVQL HQMAVIGQQA GSSSNLTELQ VVNLDTAHST KSE
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF-binding sites are also constitutive promotor elements in many muscle-specific promotors. SFR is required for cardiac differentiation and maturation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-fos serum response element-binding transcription factor; minichromosome maintenance 1 homolog; OTTHUMP00000039820; Serum response factor; serum response factor (c-fos serum response element-binding transcription factor); SRF
Gene Aliases: MCM1; SRF
UniProt ID: (Human) P11831
Entrez Gene ID: (Human) 6722
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support