Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PLDPETIAQD IEETMNTALN ELRELERQST AKHAPDVVLD TLEQVKNSPT PATSTESLSP LHNVALRSSE PQIRRSTSSS SDTMSTFKPM VAPRMGVQL
The protein encoded by this gene is a GTPase activator, working with the GTPase CDC42 to negatively regulate neuronal migration. The encoded protein interacts with the transmembrane receptor ROBO1 to inactivate CDC42.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rho GTPase activating protein 13; Rho GTPase-activating protein 13; SLIT-ROBO Rho GTPase-activating protein 1; srGAP1
Gene Aliases: 4930572H05Rik; ARHGAP13; KIAA1304; NMTC2; SRGAP1
UniProt ID: (Human) Q7Z6B7, (Mouse) Q91Z69
Entrez Gene ID: (Human) 57522, (Mouse) 117600
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support