Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAYPGHPGA GGGYYPGGYG GAPGGPAFPG QTQDPLYGYF AAVAGQDGQI DADELQRCLT QSGIAGGYKP FNLETCRLMV SMLDRDMSGT MGFNEFKELW AVLNGWRQHF ISFDTDRSGT VDPQELQKAL TTMGFRLSPQ AVNSIAKRYS TNGKITFDDY IACCVKLRAL TDSFRRRDTA QQGVVNFPYD DFIQCVMSV (1-198 aa encoded by BC011025 )
Sorcin is a 198 amino acid, soluble, resistance-related, calcium-binding cytosolic protein belonging to the penta-EF hand (PEF) family of calcium binding proteins and contains 4 EF-hand calcium-binding motifs. Human Sorcin is known to share 95% sequence identity with hamster Sorcin. It is known to associate with the sarcolemmal proteins Annexin VII, cardiac ryanodine receptors and L-type Ca2+ channels and thus influence the intracellular Ca (2+)-homeostasis. It is also important in the regulation of intracellular Ca2+ cycling and Ca2+ influx pathways in the heart and skeletal muscle during excitation-contraction. It is overproduced in many multi-drug resistance (MDR) cells and regulates cell apoptosis pathways, thus acting as a new MDR marker for prognosis of acute myeloid leukemia (AML) and a good target for anti-MDR drug development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 22 kDa protein; calcium binding protein amplified in mutlidrug-resistant cells; CP-22; CP22; H_RG167B05.1; Sorcin; V19
Gene Aliases: 2210417O06Rik; 2900070H08Rik; CP-22; CP22; SCN; Sor; SRI; V19
UniProt ID: (Human) P30626, (Mouse) Q6P069
Entrez Gene ID: (Human) 6717, (Mouse) 109552, (Rat) 683667
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support