Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MCAQYCISF ADVEKAHINI RDSIHLTPVL TSSILNQLTG RNLFFKCELF QKTGSFKIRG ALNAVRSLVP DALERKPKAV VTHSSGNHGQ ALTYAAKLEG IPAYIVVPQT APDCKKLAIQ AYGASIVYCE PSDESRENVA KRVTEETEGI MVHPNQEPAV IAGQGTIALE VLNQVPLVDA LVVPVGGGGM LAGIAITVKA LKPSVKVYAA EPSNADDCYQ SKLKGKLMPN LYPPETIADG VKSSIGLNTW PIIRDLVDDI FTVTEDEIKC ATQLVWERMK LLIEPTAGVG VAAVLSQHFQ TVSPEVKNIC IVLSGGNVDL TSSITWVKQA ERPASYQSVS V (1-340 aa encoded by BC074728)
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-serine ammonia-lyase; D-serine dehydratase; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase
Gene Aliases: ILV1; ISO1; SRR; Srs
UniProt ID: (Human) Q9GZT4, (Rat) Q76EQ0, (Mouse) Q9QZX7
Entrez Gene ID: (Human) 63826, (Rat) 303306, (Mouse) 27364
Molecular Function:
epimerase/racemase
isomerase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support