Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This intronless gene encodes a transcription factor that is a member of the high mobility group (HMG)-box family of DNA-binding proteins. This protein is the testis-determining factor (TDF), which initiates male sex determination. Mutations in this gene give rise to XY females with gonadal dysgenesis (Swyer syndrome); translocation of part of the Y chromosome containing this gene to the X chromosome causes XX male syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: essential protein for sex determination in human males; sex determining region Y protein; sex-determining region on Y; Sex-determining region Y protein; Testis-determining factor; testis-determining factor on Y
Gene Aliases: SRXX1; SRXY1; SRY; TDF; TDY
UniProt ID: (Human) Q05066
Entrez Gene ID: (Human) 6736
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support