Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: The eBioMC-813-70 monoclonal antibody reacts with a glycolipid carbohydrate epitope on Stage-specific embryonic antigen-4 (SSEA-4) protein. Expression is found on human teratocarcinoma stem cells (EC), human embryonic germ cells (EG), human induced pluripotent stem (iPS) cells, and human embryonic stem cells (ES) with a decrease in expression upon differentiation. Recently expression has also been found on bone marrow mesenchymal stem cells.
The eBioMC-813-70 antibody has been reported to crossreact to mouse, rabbit, canine, chicken, and Old and New World monkeys.
Applications Reported: This eBioMC-813-70 (MC-813-70) antibody has been reported for use in immunocytochemistry. This product has not been validated for flow cytometric analysis.
Applications Tested: This eBioMC-813-70 (MC-813-70) antibody has been tested by immunocytochemistry on fixed NTERA-2 and 2102Ep cells and can be used at less than or equal to 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable.
Excitation: 555 nm; Emission: 570 nm
SSEA-4 (Stage-specific embryonic antigen 4) is a cell surface antigen, expressed along with SSEA-3, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA-1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SSEA-4; stage-specific embryonic antigen 4
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support