Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: FYFPIPEKKE PCLQGEAESK ASKLFGNYSR DQPIFLRLED YFWVKTPSAY ELPYGTKGSE DLLLRVLAIT SSSIPKNIQS LRCRRCVVVG NGHRLRNSSL
Synthesis of alpha-2, 3-linked sialic acid to Gal(beta-1, 3)GalNAc is mediated by at least 3 distinct beta-galactoside alpha-2, 3-sialyltransferases (EC 2.4.99.4), including ST3GAL4. In contrast, only a single gene encodes the beta-galactoside alpha-2, 6-sialyltransferase, ST6GAL1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha 2,3-sialyltransferase IV; Alpha 2,3-ST 4; alpha-3-N-acetylneuraminyltransferase; beta-galactoside alpha-2,3-sialyltransferase 4; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4; FLJ11867; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase; Gal-beta-1,4-GlcNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; N-acetyllactosaminide alpha-2,3-sialyltransferase; SAT-3; Sialyltransferase 4C; sialyltransferase 4C (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4C (beta-galactoside alpha-2,3-sialytransferase); SIAT4-C; ST-4; ST3Gal IV; ST3GalA.2; ST3GalIV; STZ
Gene Aliases: CGS23; NANTA3; SAT3; SIAT4; SIAT4C; ST3GAL4; ST3GalIV; STZ
UniProt ID: (Human) Q11206
Entrez Gene ID: (Human) 6484
Molecular Function:
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support