Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LPEVYCVIS RLGCFGLFSK VLDEVERRRG ISAALVYPFM RSLMESPFPA PGKTIKVKTF LPGAGNEVLE LRRPMDSRLE HVDFECLFTC LSVRQLIRIF ASLLLERRVI FVADKLSTLS SCSHAVVALL YPFSWQHTFI PVLPASMIDI VCCPTPFLVG LLSSSLPKLK ELPVEEALMV NLGSDRFIRQ MDDEDTLLPR KLQAALEQAL ERKNELISQD SDSDSDDECN TLNGLVSEVF IRFFVETVGH YSLFLTQSEK GERAFQREAF RKSVASKSIR RFLEVFMESQ MFAGFIQDRE LRKCRAKGLF EQRVEQYLEE LPDTEQSGMN KFLRGLGNKM KFLHKKN (792-1137 aa encoded by BC036655)
This gene was identified by its ability to suppress the tumorigenicity of Hela cells in nude mice. The protein encoded by this gene contains a C-terminal region that shares similarity with the Rab 3 family of small GTP binding proteins. This protein preferentially binds to the SH3 domain of c-Abl kinase, and acts as a regulator of MAPK1/ERK2 kinase, which may contribute to its ability to reduce the tumorigenic phenotype in cells. Three alternatively spliced transcript variants of this gene encoding distinct isoforms are identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DENN domain-containing protein 2B; DENN/MADD domain containing 2B; HeLa tumor suppression 1; Suppression of tumorigenicity 5 protein
Gene Aliases: 2010004M01Rik; 2610305K15Rik; Denn2b; DENND2B; HTS1; p126; ST5
UniProt ID: (Human) P78524, (Mouse) Q924W7
Entrez Gene ID: (Human) 6764, (Mouse) 76954, (Rat) 308944
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support