Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QRAAWKSPE KEKTMVNTLS PRGQDAGMAS GRTEAQSWKS QDTKTTKGNG GQTRKLTASR TVSEKHQGKA ATTAKTLIPK SQHRMLAPTG AVSTRTRQKG VTTAVIPPKE KKPQATPPPA PFQSPTTQRN QRLKAANFKS EPRWDFEEKY SFEIGGLQTT CPDSVKIKAS KSLWLQKLFL PNLTLFLDSR HFNQSEWDRL EHFAPPFGFM ELNYSLVQKV VTRFPPVPQQ QLLLASLPAG SLRCITCAVV GNGGILNNSH IGQEIDSHDY VFRLSGALIK GYEQDVGTRT SFYGFTAFSL TQSLLILGNR GFKNVPLGKA QTPGSFSGSP AHGQVPVAAP RLSPIHEEQV SEV (120-471 aa encoded by BC022462)
Glycosylation of proteins affects cell-cell interaction, interactions with the matrix, and the functions of intracellular molecules. ST6GALNAC1 transfers a sialic acid, N-acetylneuraminic acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; GalNAc alpha-2, 6-sialyltransferase I, long form; GalNAc alpha-2,6-sialyltransferase I; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-beta-galactosyl-1,3)-N-acetyl galactosaminde alpha-2,6-sialyltransferase) A; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) A; Sialyltransferase 7A; SIAT7-A; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; ST6 GalNAc alpha-2,6-sialyltransferase 1; ST6GalNAc I; ST6GalNAc-I; ST6GALNAC1, alpha-2,6-sialyltransferase 1; ST6GalNAcI
Gene Aliases: HSY11339; SIAT7A; ST6GALNAC1; ST6GalNAcI; STYI; UNQ543/PRO848
UniProt ID: (Human) Q9NSC7, (Mouse) Q9QZ39
Entrez Gene ID: (Human) 55808, (Mouse) 20445
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support