Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: Tonsil
Cellular location: Nucleus and cytoplasm.
The protein encoded by this gene functions as a docking protein acting downstream of Tec tyrosine kinase in B cell antigen receptor signaling. The protein is directly phosphorylated by Tec in vitro where it participates in a postive feedback loop, increasing Tec activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCR downstream signaling 1; BCR downstream-signaling protein 1; Docking protein BRDG1; Signal-transducing adaptor protein 1; STAP-1; Stem cell adaptor protein 1; stem cell adaptor protein STAP-1
Gene Aliases: AI586015; BRDG1; RGD1311584; STAP-1; STAP1
UniProt ID: (Human) Q9ULZ2, (Mouse) Q9JM90
Entrez Gene ID: (Human) 26228, (Rat) 305269, (Mouse) 56792
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support