Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ESQFSVGSN ELVFQVKTLS LPVVVIVHGS QDHNATATVL WDNAFAEPGR VPFAVPDKVL WPQLCEALNM KFKAEVQSNR GLTKENLVFL AQKLFNNSSS HLEDYSGLSV SWSQFNRENL PGWNYTFWQW FDGVMEVLKK HHKPHWNDGA ILGFVNKQQA HDLLINKPDG TFLLRFSDSE IGGITIAWKF DSPERNLWNL KPFTTRDFSI RSLADRLGDL SYLIYVFPDR PKDEVFSKYY TPVLAKAVDG YVKPQIKQVV PEFVNASADA GGSSATYMDQ APSPAVCPQA PYNMYPQNPD HVLDQDGEFD LDETMDVARH VEELLRRPMD SLDSRLSPPA GLFTSARGSL S (445-794 aa encoded by BC027036)
STAT5 alpha is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. STAT5 alpha is activated by, and mediates the responses of many cell ligands, such as IL2, IL3, IL7 GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of STAT5 alpha in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of STAT5 alpha is found to induce the expression of BCL2L1/BCL-X(L), which suggests the antiapoptotic function of this protein in cells. STAT5 alpha, along with STAT5 beta, were identified in mouse. The amino acid sequences of STAT5 alpha and STAT5 beta show 96% sequence similarity, and both proteins are co expressed in most tissues in virgin and lactating mice. However, differential accumulation of STAT5 alpha and STAT5 beta mRNA has been reported for both muscle and mammary tissue. STAT5 alpha is critically involved in a variety of physiological functions, including reproduction, lactation, immune function and somatic growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mammary gland factor; mammary gland factor STAT5A; mammary gland factor; transcription factor; signal transducer and activator of transcription 5; Signal transducer and activator of transcription 5A; STAT; STAT5B
Gene Aliases: AA959963; MGF; Mpf; STAT5; STAT5A
UniProt ID: (Human) P42229, (Pig) Q9TUZ1, (Rat) Q62771, (Mouse) P42230
Entrez Gene ID: (Human) 6776, (Pig) 397550, (Rat) 24918, (Mouse) 20850
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support