Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MEVVDPQQLG MFTEGELMSV GMDTFIHRID STEVIYQPRR KRAKLIGKYL MGDLLGEGSY GKVKEVLDSE TLCRRAVKIL KKKKLRRIPN GEANVKKEIQ LLRRLRHKNV IQLVDVLYNE EKQKMYMVME YCVCGMQEML DSVPEKRFPV CQAHGYFCQL IDGLEYLHSQ GIVHKDIKPG NLLLTTGGTL KISDLGVAEA LHPFAADDTC RTSQGSPAFQ PPEIANGLDT FSGFKVDIWS AGVTLYNITT GLYPFEGDNI YKLFENIGKG SYAIPGDCGP PLSDLLKGML EYEPAKRFSI RQIRQHSWFR KKHPPAEAPV PIPPSPDTKD RWRSMTVVPY LEDLHGADED EDLFDIEDDI IYTQDFTVPG QVPEEEASHN GQRRGLPKAV CMNGTEAAQL STKSRAEGRA PNPARKACSA SSKIRRLSAC KQQ
LKB1 (also known as STK11 - Serine/threonine-protein kinase STK11), a member of the serine/threonine kinase family, is a tumor suppressor. LKB1 physically associates with p53 and regulates specific p53 dependent apoptosis pathways. LKB1 is present in both the cytoplasm and nucleus of living cells and translocates to mitochondria during apoptosis. LKB1 functions as a master upstream protein kinase, regulating AMPK related kinases as well as AMPK. Mutations in LKB1 gene are associated with Peutz Jeghers syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAMK group; CAMKL family; Liver kinase B1; LKB 1; LKB-1; LKB1; LKB1(S); polarization-related protein LKB1; Renal carcinoma antigen NY-REN-19; serine/threonine-protein kinase 11; serine/threonine-protein kinase LKB1; Serine/threonine-protein kinase STK11
Gene Aliases: hLKB1; LKB1; PJS; STK11
UniProt ID: (Human) Q15831
Entrez Gene ID: (Human) 6794
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support