Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MGHALCVCSR GTVIIDNKRY LFIQKLGEGG FSYVDLVEGL HDGHFYALKR ILCHEQQDRE EAQREADMHR LFNHPNILRL VAYCLRERGA KHEAWLLLPF
STK16 (PKL12) is a serine/threonine kinase that functions in signal transduction pathways and transcriptional regulation. Protein kinase that act on both serine and threonine residues. STK16 may regulate transforming growth factor-beta (Tgfb) and vascular endothelial growth factor transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPSK; MPSK; Myristoylated and palmitoylated serine/threonine-protein kinase; protein kinase expressed in day 12 fetal liver; Protein kinase PKL12; Serine/threonine-protein kinase 16; TGF-beta-stimulated factor 1; TSF-1; Tyrosine-protein kinase STK16
Gene Aliases: KRCT; MPSK; MPSK1; PKL12; STK16; TSF1
UniProt ID: (Human) O75716
Entrez Gene ID: (Human) 8576
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support