Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MIPLEKPGS GGSSPGATSG SGRAGRGLSG PCRPPPPPQA RGLLTEIRAV VRTEPFQDGY SLCPGRELGR GKFAVVRKCI KKDSGKEFAA KFMRKRRKGQ DCRMEIIHEI AVLELAQDNP WVINLHEVYE TASEMILVLE YAAGGEIFDQ CVADREEAFK EKDVQRLMRQ ILEGVHFLHT RDVVHLDLKP QNILLTSESP LGDIKIVDFG LSRILKNSEE LREIMGTPEY VAPEILSYDP ISMATDMWSI GVLTYVMLTG ISPFLGNDKQ ETFLNISQMN LSYSEEEFDV LSESAVDFIR TLLVKKPEDR ATAEECLKHP WLTQSSI (1-326 aa encoded by BC047696)
STK17A (DRAK1) is a serine/threonine kinase localized in the nucleus and involved in apoptosis. Diseases associated with STK17A include Colon Squamous Cell Carcinoma. Among its related pathways are Sweet Taste Signaling. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is STK17B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAP kinase-related apoptosis-inducing protein kinase 1; death-associated protein kinase-related 1; serine/threonine kinase 17a (apoptosis-inducing); Serine/threonine-protein kinase 17A
Gene Aliases: DRAK1; STK17A
UniProt ID: (Human) Q9UEE5
Entrez Gene ID: (Human) 9263
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support