Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to STK38L.
Immunogen sequence: FESLKVIGR GAFGEVRLVQ KKDTGHIYAM KILRKSDMLE KEQVAHIRAE RDILVEADGA WVVKMFYSFQ DKRNLYLIME FLPGGDMMTL LMKKDTLTEE ETQFYISETV LAIDAIHQLG FIHRDIKPDN LLLDAKGHVK LSDFGLCTGL KKAHRTEFYR NLTHNPPSDF SFQNMNSKRK AETWKKNRRQ LAYSTVGTPD YIAPEVFMQT GYNKLCDWWS LGVIMYEMLI GYPPFCSETP QETYRKVMNW KETLVFPPEV PISEKAKDLI LRFCIDSENR IGNSGVEE (90-376 aa encoded by BC028603 )
STK38L (also known as NDR2) or Nuclear Dbf2-related protein is a serine/threonine kinase in the subgroup of the AGC kinase family. STK38L act as novel pro-apoptotic kinase that is activated by RASSF1A/MST1 in response to Fas receptor stimulation and promotes apoptosis. STK38L is mainly involved in the regulation of structural processes in differentiating and mature neuronal cells and it is highly expressed with highest levels observed in the thymus. STK38L contains 12 protein kinase catalytic subdomains and a potential bipartite nuclear localization signal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NDR2 protein kinase; nuclear Dbf2-related 2; Nuclear Dbf2-related kinase 2; serine/threonine kinase 38-like; Serine/threonine-protein kinase 38-like; STK38L
Gene Aliases: 4930473A22Rik; B230328I19; KIAA0965; NDR2; Ndr54; RGD1564793; STK38L
UniProt ID: (Human) Q9Y2H1, (Mouse) Q7TSE6
Entrez Gene ID: (Human) 23012, (Mouse) 232533, (Rat) 691337
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support