Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
STK40 is a protein coding gene. May be a negative regulator of NF-kappa-B and p53-mediated gene transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ortholog of Ser/Thr-like protein kinase Lyk4; RP11-268J15.4; Ser/Thr-like kinase; Ser/Thr-like protein kinase lyk4; Serine/threonine-protein kinase 40; Serine/threonine-protein kinase lyk4; SgK495; SINK-homologous serine/threonine-protein kinase; Sugen kinase 495
Gene Aliases: 2310004N11Rik; AI428792; Lyk4; RGD1307310; SGK495; SHIK; STK40
UniProt ID: (Human) Q8N2I9, (Rat) Q7TNL4, (Mouse) Q7TNL3
Entrez Gene ID: (Human) 83931, (Rat) 360230, (Mouse) 74178
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support