Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVATWRVLL SALYNAIHLG QMDLSLLPPR AATLDPGYYT YRNFLKIEVS QSHPAMTAFC SLLLQAQSLL PRTMAAPQDS LRPGEEDEGM QLLQTKDSMA KGARPGASRG RARWGLAYTL LHNPTLQVFR KTALLGANGA QP (527-667 aa encoded by BC025256)
STRA6 is a membrane protein involved in the metabolism of retinol. The encoded protein acts as a receptor for retinol/retinol binding protein complexes. This protein removes the retinol from the complex and transports it across the cell membrane. Defects in this gene are a cause of syndromic microphthalmia type 9 (MCOPS9).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ12541; Receptor for retinol uptake STRA6; Retinoic acid-responsive protein; retinol binding protein 4 receptor; Retinol-binding protein receptor STRA6; stimulated by retinoic acid 6 homolog; stimulated by retinoic acid gene 6 homolog; Stimulated by retinoic acid gene 6 protein; Stimulated by retinoic acid gene 6 protein homolog
Gene Aliases: AI891933; MCOPCB8; MCOPS9; PP14296; STRA6; UNQ3126/PRO10282/PRO19578
UniProt ID: (Human) Q9BX79, (Mouse) O70491
Entrez Gene ID: (Human) 64220, (Mouse) 20897
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support