Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSQPAGNQT SPGATEDYSY GSWYIDEPQG GEELQPEGEV PSCHTSIPPG LYHACLASLS ILVLLLLAML VRRRQLWPDC VRGRPGLPSP VDFLAGDRPR AVPAAVFMVL LSSLCLLLPD EDALPFLTLA SAPSQDGKTE APRGAWKILG LFYYAALYYP LAACATAGHT AAHLLGSTLS WAHLGVQVWQ RAECPQVPKI YKYYSLLASL PLLLGLGFLS LWYPVQLVRS FSRRTGAGSK GLQSSYSEEY LRNLLCRKKL GSSYHTSKHG FLSWARVCLR HCIYTPQPGF HLPLKLVLSA TLTGTAIYQV ALLLLVGVVP TIQKVRAGVT TDVSYLLASF GIVLSEDKQE VVELVKHHLW ALEVCYISAL VLSCLLTFLV LMRSLVTHRT NLRALHRGAA LDLSPLHRSP HPSRQAIFCW MSFSAYQTAF ICLGLLVQQI IFFLGTTALA FLVLMPVLHG RNLLLFRSLE SSWPFWLTLA LAVILQNMAA HWVFLETHDG HPQLTNRRVL YAATFLLFPL NVLVGAMVAT WRVLLSALYN AIHLGQMDLS LLPPRAATLD PGYYTYRNFL KIEVSQSHPA MTAFCSLLLQ AQSLLPRTMA APQDSLRPGE EDEGMQLLQT KDSMAKGARP GASRGRARWG LAYTLLHNPT LQVFRKTALL GANGAQP
STRA6 is a membrane protein involved in the metabolism of retinol. The encoded protein acts as a receptor for retinol/retinol binding protein complexes. This protein removes the retinol from the complex and transports it across the cell membrane. Defects in this gene are a cause of syndromic microphthalmia type 9 (MCOPS9).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ12541; Receptor for retinol uptake STRA6; retinol binding protein 4 receptor; Retinol-binding protein receptor STRA6; stimulated by retinoic acid 6 homolog; stimulated by retinoic acid gene 6 homolog; Stimulated by retinoic acid gene 6 protein homolog
Gene Aliases: MCOPCB8; MCOPS9; PP14296; STRA6; UNQ3126/PRO10282/PRO19578
UniProt ID: (Human) Q9BX78
Entrez Gene ID: (Human) 64220
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support