Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
STRAP is a 350 amino acid MAP activator with WD-40 repeats that is localized both in the cytoplasm and the nucleus. It is a ubiquitously expressed protein that acts as an inhibitor of TGF-beta signaling and an important regulator of cell proliferation. Stable expression of STRAP results in activation of the mitogen-activated protein kinase/extracellular signal-regulated kinase pathway and in down-regulation of the cyclin-dependent kinase inhibitor p21 (Cip1), which results in retinoblastoma protein hyperphosphorylation. Upregulation of STRAP might play a role in tumor development as an early event for colorectal cancers. It is the first component of the U snRNP assembly machinery that associates with SMN complex in a compartment-specific way and plays a crucial role in the intracellular distribution of SMN1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAP activator with WD repeats; Serine-threonine kinase receptor-associated protein; UNR-interacting protein; WD-40 repeat protein PT-WD
Gene Aliases: AW557906; C78091; C79202; MAWD; PT-WD; STRAP; UNRIP
UniProt ID: (Human) Q9Y3F4, (Mouse) Q9Z1Z2
Entrez Gene ID: (Human) 11171, (Mouse) 20901
Molecular Function:
translation initiation factor
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support