Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SNSEPNGSV ETKNLEQILN GGESPKQKGQ EIKRSSGDVL ETFNFLENAD DSDEDEENDM IEGIPEGKDK HRMNKHKIGN EGLAADLTDD PDTEE (207-300 aa encoded by BC126221)
SG2NA was identified as a human cancer autoantigen and has now been implicated in vesicular trafficking and signal transduction. It is highly related to striatin and zinedin. It is a scaffolding protein that binds PP2A A and C subunits as well as the human homolog of the yeast protein, Mob1, which has been implicated in control of mitotic progression in yeast. It also binds calmodulin and appears to modulate PP2A activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell cycle autoantigen SG2NA; cell cycle S/G2 nuclear autoantigen; cell-cycle autoantigen SG2NA; nuclear autoantigen; protein phosphatase 2 regulatory subunit B'''beta; S/G2 antigen; SG2NA beta isoform; striatin, calmodulin binding protein 3; Striatin-3; striatin-3 35 kDa; striatin-3-gamma
Gene Aliases: GS2NA; PPP2R6B; SG2NA; STRN3
UniProt ID: (Human) Q13033, (Mouse) Q9ERG2, (Rat) P58405
Entrez Gene ID: (Human) 29966, (Mouse) 94186, (Rat) 114520
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support