Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KERLMNDFSAA LNNFQAVQRR VSEKEKESIA RARAGSRLSA EERQREEQLV SFDSHEEWNQ MQSQEDEVAI TEQDLELIKE RET
Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC51957; Syntaxin-12
Gene Aliases: STX12; STX13; STX14
UniProt ID: (Human) Q86Y82
Entrez Gene ID: (Human) 23673
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support