Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-26001 detects a band of ~38 kDa corresponding to recombinant rat syntaxin 13. Suggested positive controls include rat or mouse brain extract.
SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC51957; Syn13; syntaxin 12; syntaxin 13; Syntaxin-12; Syntaxin-13
Gene Aliases: AI850350; AU041521; Stx12; Stx13
UniProt ID: (Rat) G3V7P1, (Mouse) Q9ER00
Entrez Gene ID: (Rat) 65033, (Chinese hamster) 100773207, (Mouse) 100226
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support