Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: QIFMRTCSEA IQQLRTEAHK EIHSQQVKEH RTAVLDFIED YLKRVCKLYS EQRAIRVKRV VDKKRLSKLE PEPNTKTRES TSSEKVSQSP SKDSEENPA
Syntaxins, a family of proteins involved in the fusion of synaptic vesicles with the plasma membrane, display broad tissue distribution and contain carboxy-terminal hydrophobic domains that direct themselves to their respective intracellular compartments. Syntaxin 18 mainly localizes to the endoplasmic reticulum (ER), and functions in transport between the ER and Golgi. Syntaxin 18 also plays a role in ER-mediated phagocytosis, possibly by regulating the specific fusion of the ER and plasma or phagosomal membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell growth-inhibiting gene 9 protein; Syntaxin-18
Gene Aliases: 1810035L21Rik; 4933425D03Rik; AU041562; GIG9; STX18; Ufe1
UniProt ID: (Human) Q9P2W9, (Mouse) Q8VDS8, (Rat) Q68FW4
Entrez Gene ID: (Human) 53407, (Mouse) 71116, (Rat) 360953
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support