Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MPGLTCFTHD NQHWQTAPFW TLGPFCACTS ANNNTYWCMR TINETHNFLF CEFATGFLEY FDLNTDPYQL MNAVNTLDRD VLNQLHVQLM ELRSCKGYKQ CNPRTRNMDL GLKDGGSYEQ YRQFQRRKWP EMKRPSSKSL GQLWEGWEG
Heparan sulfate 6-O-endosulfatases, such as Sulfatase 2, selectively remove 6-O-sulfate groups from heparan sulfate. Heparan sulfate proteoglycans (HSPGs) act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules (Dai et al., 2005 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arylsulfatase; DKFZp313E091; Extracellular sulfatase Sulf-2; FLJ44488; FLJ90554; hSulf-2; MGC126411; N-acetylglucosamine-6-sulfatase; OTTHUMP00000031223; RP5-1049G16.1
Gene Aliases: HSULF-2; KIAA1247; SULF2; UNQ559/PRO1120
UniProt ID: (Human) Q8IWU5
Entrez Gene ID: (Human) 55959
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support