Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAESEAETP STPGEFESKY FEFHGVRLPP FCRGKMEEIA NFPVRPSDVW IVTYPKSGTS LLQEVVYLVS QGADPDEIGL MNIDEQLPVL EYPQPGLDII KELTSPRLIK SHLPYRFLPS DLHNGDSKVI YMARNPKDLV VSYYQFHRSL RTMSYRGTFQ EFCRRFMNDK LGYGSWFEHV QEFWEHRMDS NVLFLKYEDM HRDLVTMVEQ LARFLGVSCD KAQLEALTEH CHQLVDQCCS AEALPVGRGR VGLWKDIFTV SMNEKFDLVY KQKMGKCDLT FDFYL (1-284 aa encoded by BC022459)
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure(number and length of exons) is similar among family members. SULT4A1 isoforms a and b are selectively expressed in brain tissue. Isoform a is the predominant of the two isoforms of this protein. Isoform b includes an alternate segment, compared to isoform a, that causes a frameshift. The resulting protein (isoform b) has a distinct and shorter C-terminus when compared to isoform a.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain sulfotransferase-like protein; hBR-STL; mBR-STL; nervous system cytosolic sulfotransferase; Nervous system sulfotransferase; NST; ST4A1; Sulfotransferase 4A1; sulfotransferase family 4A, member 1; sulfotransferase-related protein
Gene Aliases: 2400007A17Rik; AI853543; BR-STL-1; BRSTL1; DJ388M5.3; hBR-STL-1; NST; ST4A1; SULT4A1; SULTX3
UniProt ID: (Human) Q9BR01, (Mouse) P63046
Entrez Gene ID: (Human) 25830, (Mouse) 29859
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support