Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MGTLLGLGAV LAYQDHRCRA AQESTHIYTK EEVSSHTSPE TGIWVTLGSE VFDVTEFVDL HPGGPSKLML AAGGPLEPFW ALYAVHNQSH VRELLAQYKI GELNPEDKVA PTVETSDPYA DDPVRHPALK VNSQRPFNAE PPPELLTENY ITPNPIFFTR NHLPVPNLDP DTYRLHVVGA PGGQSLSLSL DDLHNFPRYE ITVTLQCAGN RRSEMTQVKE VKGLEWRTGA ISTARWAGAR LCDVLAQAGH QLCETEAHVC FEGLDSDPTG TAYGASIPLA RAMDPEAEVL LAYEMNGQPL PRDHGFPVRV VVPGVVGARH VKWLGRVSVQ PEESYSHWQR RDYKGFSPSV DWETVDFDSA PSIQELPVQS AITEPRDGET VESGEVTIKG YAWSGGGRAV IRVDVSLDGG LTWQVAKLDG EEQRPRKAWA WRLWQLKAPV PAGQKELNIV CKAVDDGYNV QPDTVAPIWN LRGVLSNAWH RVHVYVSP
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sulfite oxidase, mitochondrial
Gene Aliases: SUOX
UniProt ID: (Human) P51687
Entrez Gene ID: (Human) 6821
Molecular Function:
oxidoreductase
reductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support