Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GCSVCCKSS WNQLQDLCRL AKLSCPALGI SKRNLYDFEV EYLCDYKKIR EQEYYLVKWR GYPDSESTWE PRQNLKCVRI LKQFHKDLER ELLRRHHRSK TPRHLDPSLA NYLVQKAKQR RALRRWEQEL NAKRSHLGRI TVENEVDLDG PPRAFVYINE YRVGEGITLN QVAVGCECQD CLWAPTGGCC PGASLHKFAY NDQGQVRLRA GLPIYECNSR CRCGYDCPNR VVQKGIRYDL CIFRTDDGRG WGVRTLEKIR KNSFVM (7-271 aa encoded by B C006238)
This gene encodes an evolutionarily-conserved protein containing an N-terminal chromodomain and a C-terminal SET domain. The encoded protein is a histone methyltransferase that trimethylates lysine 9 of histone H3, which results in transcriptional gene silencing. Loss of function of this gene disrupts heterochromatin formation and may cause chromosome instability. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H3-K9-HMTase 1; Histone H3-K9 methyltransferase 1; Histone-lysine N-methyltransferase SUV39H1; histone-lysine N-methyltransferase, H3 lysine-9 specific 1; Lysine N-methyltransferase 1A; Position-effect variegation 3-9 homolog; Su(var)3-9 homolog 1; Suppressor of variegation 3-9 homolog 1; Suv39h histone methyltransferase
Gene Aliases: AI852103; AL022883; DXHXS7466e; H3-K9-HMTase 1; KMT1A; MG44; mIS6; RGD1565028; SUV39H; SUV39H1
UniProt ID: (Human) O43463, (Mouse) Q3TEW2
Entrez Gene ID: (Human) 6839, (Mouse) 20937, (Rat) 302553
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support