Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEYYLVKWKG WPDSTNTWEP LQNLKCPLLL QQFSNDKHNY LSQVKKGKAI TPKDNNKTLK PAIAEYIVKK AKQRIALQRW QDELNRRKNH KGMIFVENTV DLEGPPSDFY YINEYKPAPG ISLVNEATFG CSCTDCFFQK CCPAEAGVLL AYNKNQQIKI PPGTPIYECN SRCQCGPDCP NRIVQKGTQY SLCIFRTSNG RGWGVKTLVK IKRMSFVMEY VGEVITSEEA ERRGQFYDNK GITYLFDLDY ESDEFTVDAA RYGNVSHFVN HSCDPNLQVF NVFIDNLDTR LPRIALFSTR TINAGEELTF DYQMKGSGDI SSDSIDHSPA KKRVRTVCKC GAVTCRGYLN
The murine gene Suv39h2 encodes an H3 histone methyltransferase (HMTase) 59% identical in sequence to mouse Suv39h1. During embryogenesis, both proteins overlap in tissue expression, yet Suv39h2 transcripts are restricted to the testes in adult animals. Immunolocalization of the Suv39h2 protein during spermatogenesis indicates enrichment at the heterochromatin from the leptotene to the round spermatid stage. Moreover, Suv39h2 specifically accumulates with chromatin of the sex chromosomes, which undergo transcriptional silencing during the first meiotic prophase. Suv39h2 HMTase may also organize meiotic heterochromatin with the potential for epigenetic imprint to the male germline.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ23414; H3 K9 HMTase 2; H3-K9-HMTase 2; Histone H3-K9 methyltransferase 2; Histone-lysine N-methyltransferase SUV39H2; KMT1; Lysine N-methyltransferase 1B; RP11-2K17.2; Su; Su(var)3-9 homolog 2; Suppressor of variegation 3-9 homolog 2
Gene Aliases: KMT1B; SUV39H2
UniProt ID: (Human) Q9H5I1
Entrez Gene ID: (Human) 79723
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support