Search Thermo Fisher Scientific
FIGURE: 1 / 1
The SV40 T antigen is encoded by the early region of the SV40 genome. The large T antigen binds DNA, and complexes with a 53,000 dalton cellular protein, p53, which is required for initiation of viral DNA replication during lytic growth. In addition, the large T antigen binds DNA polymerase and the transcription factor AP-2 and forms a specific complex with the P105 product of the retinoblastoma susceptibility gene.
Protein Aliases: simian vacuolating virus 40; simian virus; Simian virus 40; sv40 large t antigen; sv40 virus large t antigen