Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DLDMQKAEEQ EEKILNMFRQ QQKILQQSRI VQSQRLKTIK QLYEQFIKSM EELEKNHDNL LTGAQNEFKK EMAMLQKKIM METQQQEIAS VRKSLQSMLF
This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregnancy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SCP-3; Synaptonemal complex protein 3; synaptonemal complex protein 3-like
Gene Aliases: COR1; RNASCP3; RPRGL4; SCP3; SPGF4; SYCP3
UniProt ID: (Human) Q8IZU3, (Rat) Q63520, (Mouse) P70281
Entrez Gene ID: (Human) 50511, (Rat) 25561, (Mouse) 20962
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support