Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: GEESTSPEQA QTLGQDSLGP PEHFQGGPRG NEPAAHPPRW STPSSYEDPA
Sequence homology: Cow: 85%; Horse: 85%; Human: 100%
The exact function of C19orf46 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: deafness, autosomal recessive 76; KASH domain-containing protein 4; KASH4; Nesprin-4; Nuclear envelope spectrin repeat protein 4; spectrin repeat containing, nuclear envelope family member 4
Gene Aliases: C19orf46; DFNB76; Nesp4; SYNE4
UniProt ID: (Human) A8MRS0
Entrez Gene ID: (Human) 163183
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support