Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: SPPSYSVLYP SSDPKSSHLK GQAVPASKTG ILEESMARRG SRKSMFTFVE KPKVTPNPDL LDLVQTADEK RRQRDQGEVG VEEEPFALGA EASNFQQEPA PRDRASPAAA EEVVPEWASC LKSPRIQAKP KPKPNQNLSE ASGKGAELYA RRQSRMEKYV IESSSHTPEL ARCPSPTMSL PSSWKYPTNA PGAFRVASRS PARTPPASLY HGYLPENGVL RPEPTKQPPY QLRPSLFVLS PIKEPAKVSP RAASPAKPSS LDLVPNLPKG ALPPSPALPR PSRSSPGLYT SPGQDSLQPT AVSPPYGGDI SPVSPSRAWS PRAKQAPRPS FSTRNAGIEA QVWKPSFCFK
SYNPO (Synaptopodin) is an actin-associated protein in telencephalic dendrites and renal podocytes. SYNPO is tightly associated with the dendritic spine apparatus, and mice lacking the SYNPO gene lack the apparatus. These SYNPO-null mice also demonstrate an impaired ability to express long-term potentiation as well as deficits in spatial memory tasks, indicating that SYNPO is involved in the regulation of synaptic plasticity. Recent studies suggest that SYNPO is linked to neuronal calcium stores and plays a role in the calcium store-associated ability of neurons to undergo long-term plasticity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Synaptopodin
Gene Aliases: 9030217H17Rik; 9130229N11; 9330140I15Rik; AW046661; KIAA1029; SYNPO
UniProt ID: (Human) Q8N3V7, (Rat) Q9Z327
Entrez Gene ID: (Human) 11346, (Pig) 110259543, (Rat) 60324, (Mouse) 104027
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support