Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MVSESHHEAL AAPPVTTVAT VLPSNATEPA SPGEGKEDAF SKLKEKFMNE LHKIPLPPWA LIAIAIVAVL LVLTCCFCIC KKCLFKKKNK KKGKEKGGKN AINMKDVKDL GKTMKDQALK DDDAETGLTD GEEKEEPKEE EKLGKLQYSL DYDFQNNQLL VGIIQAAELP ALDMGGTSDP YVKVFLLPDK KKKFETKVHR KTLNPVFNEQ FTFKVPYSEL GGKTLVMAVY DFDRFSKHDI IGEFKVPMNT VDFGHVTEEW RDLQSAEKEE QEKLGDICFS LRYVPTAGKL TVVILEAKNL KKMDVGGLSD PYVKIHLMQN GKRLKKKKTT IKKNTLNPYY NESFSFEVPF EQIQKVQVVV TVLDYDKIGK NDAIGKVFVG YNSTGAELRH WSDMLANPRR PIAQWHTLQV EEEVDAMLAV KK
Synaptotagmin 1 is a synaptic vesicle membrane glycoprotein that is widely expressed throughout the CNS and is generally thought to act as the Ca2+ sensor in the regulation of exocytosis and neurotransmitter release. Studies indicate that synaptotagmin’s Ca2+ mediated binding of SNAP25 is essential for the Ca2+ dependent triggering of membrane. It has been demonstrated that discrete residues within the c(2)b binding domain of synaptotagmin 1 independently specify endocytic rate and synaptic vesicle size. Synaptotagmin 1 may have a regulatory role in the membrane interactions during trafficking of synaptic vesicles at the active zone of the synapse. Further, Synaptotagmin 1 binds acidic phospholipids with a specificity that requires the presence of both an acidic head group and a diacyl backbone. A Ca(2+)-dependent interaction between synaptotagmin and putative receptors for activated protein kinase C has been reported. Synaptotagmin 1 can also bind to at least three additional proteins in a Ca(2+)-independent manner; these are neurexins, syntaxin and AP2. Diseases associated with the dysfunction of Synaptotagmin 1 include food-borne botulism and infant botulism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781D2042; FLJ42519; p65; SV DKFZp781D2042; SV SYT; Syb1; synaptotagmin 2; Synaptotagmin I; Synaptotagmin-1; SYT2; SytI
Gene Aliases: P65; SVP65; SYT; SYT1
UniProt ID: (Human) P21579
Entrez Gene ID: (Human) 6857
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support