Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IGSMMSIYS EAGDFGNIFV TGRIAFSLKY EQQTQSLVVH VKECHQLAYA DEAKKRSNPY VKTYLLPDKS RQGKRKTSIK RDTVNPLYDE TLRYEIPESL LAQRTLQFSV WHHGRFGRNT FLGEAEIQMD SWKLDKKLDH CLPLHGKISA ESPTGLPSHK GELVVSLKYI PASKTPVGGD RKKSKGGEGG ELQVWIKEAK NLTAAKAGGT SDSFVKGYLL PMRNKASKRK TPVMKKTLNP HYNHTFVYNG VRLEDLQHMC LELTVWDREP LASNDFLGGV RLGVGTGISN GEVVDWMDST GEEVSLWQKM RQYPGSWAEG TLQLRSSMAK QKLGL (338-671 aa encoded by BC014913)
This gene encodes a member of the synaptotagmin like protein family. Members of this family are characterized by an N-terminal Rab27 binding domain and C-terminal tandem C2 domains. The encoded protein binds specific small Rab GTPases and is involved in intracellular membrane trafficking. This protein binds Rab27 and may be involved in inhibiting dense core vesicle exocytosis. Alternate splicing results in multiple transcript variants that encode the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exophilin-2; Granuphilin; granuphilin b; granuphilin-a; granuphilin-b; synaptotagmin-like 4 (granuphilin-a); Synaptotagmin-like protein 4
Gene Aliases: Gph; SLP4; SYTL4
UniProt ID: (Human) Q96C24, (Mouse) Q9R0Q1, (Rat) Q8VHQ7
Entrez Gene ID: (Human) 94121, (Mouse) 27359, (Rat) 140594
Molecular Function:
myelin protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support