Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
This antibody has been tested in direct-ELISA
Homocysteine S-methyltransferase involved in the conversion of S-adenosylmethionine (AdoMet) to methionine to control the methionine/AdoMet ratio. Converts also S-methylmethionine (SMM) to methionine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hcy S-methyltransferase 1; homocysteine methyltransferase 1; Homocysteine S-methyltransferase 1; S-methylmethionine; S-methylmethionine:homocysteine methyltransferase 1; SMM; SMM:Hcy S-methyltransferase 1
Gene Aliases: L0552; MHT1; YLL062C
UniProt ID: (Baker's yeast) Q12525
Entrez Gene ID: (Baker's yeast) 850664
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support