Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
This antibody has been tested in direct-ELISA
Responsible for the proteolytic maturation of the alpha-factor precursor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dipeptidyl aminopeptidase A; DPAP A; YCI1; YOR219C; YSCIV
Gene Aliases: STE13; YCI1; YOR219C; YOR50-9
UniProt ID: (Baker's yeast) P33894
Entrez Gene ID: (Baker's yeast) 854394
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support