Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 150 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: Salmonella (ENZ15).
The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major surface antigens; the H or flagellar antigen (phase 1 and 2), the O or somatic antigen (part of the LPS moiety) and the Vi or capsular antigen (referred to as K in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram negative bacteria. This LPS is composed of an O polysaccharide (O antigen) an R core and the endotoxic inner Lipid A. Endotoxins evoke fever and can activate complement, kinin and clotting factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Salmonella
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support