Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Drosophila melanogaster saxophone (sax) gene, a previously identified female sterile locus, encodes a TGF-beta-like type I receptor. Embryos from sax mothers and dpp embryos exhibit similar mutant phenotypes during early gastrulation, and these two loci exhibit genetic interactions, which suggest that they are utilized in the same pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG1891; CG1891-PA; CG1891-PB; CG1891-PC; GH13369p; Receptor protein serine/threonine kinase; sax; sax-PA; sax-PB; sax-PC
Gene Aliases: Bkr43E; Brk43E; CG1891; Dmel\CG1891; Dmel_CG1891; Sa; SAX; STK-B
Entrez Gene ID: (Fruit fly) 35731
Molecular Function:
serine/threonine protein kinase receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support