Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKPNIIFVL SLLLILEKQA AVMGQKGGSK GRLPSEFSQF PHGQKGQHYS GQKGKQQTES KGSFSIQYTY HVDANDHDQS RKSQQYDLNA LHKTTKSQRH LGGSQQLLHN KQEGRDHDKS KGHFHRVVIH HKGGKAHRGT QNPSQDQGNS PSGKGISSQY SNTEERLWVH GLSKEQTSVS GAQKGRKQGG SQSSYVLQTE ELVANKQQRE TKNSHQNKGH YQNVVEVREE HSSKVQTSLC PAHQDKLQHG SKDIFSTQDE LLVYNKNQHQ TKNLNQDQQH GRKANKISYQ SSSTEERRLH YGENGVQKDV SQR (1-312 aa encoded by BC007096)
The protein encoded by this gene is the predominant protein in semen. The encoded secreted protein is involved in the formation of a gel matrix that encases ejaculated spermatozoa. The prostate-specific antigen (PSA) protease processes this protein into smaller peptides, with each possibly having a separate function. The proteolysis process breaks down the gel matrix and allows the spermatozoa to move more freely.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 103; MGC14719; semen coagulating protein; Semenogelin I; Semenogelin-1; SGI; SgI-29; Svp-1; Svp5; Svs5
Gene Aliases: CT103; dJ172H20.2; SEMG; SEMG1; SGI
UniProt ID: (Human) P04279
Entrez Gene ID: (Human) 6406
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support