Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Samples: 293T
Immunogen sequence: MNKPLLWISV LTSLLEAFAH TDLSGKVFVF PRESVTDHVN LITPLEKPLQ NFTLCFRAYS DLSRAYSLFS YNTQGRDNEL LVYKERVGEY SLYIGRHKVT SKVIEKFPAP VHICVSWESS SGIAEFWING TPLVKKGLRQ GYFVEAQPKI VLGQEQDSYG GKFDRSQSFV GEIGDLYMWD SVLPPENILS AYQGTPLPAN ILDWQALNYE IRGYVIIKPL VWV
The protein encoded by this gene is a glycoprotein, belonging to the pentraxin family of proteins, which has a characteristic pentameric organization. These family members have considerable sequence homology which is thought to be the result of gene duplication. The binding of the encoded protein to proteins in the pathological amyloid cross-beta fold suggests its possible role as a chaperone. This protein is also thought to control the degradation of chromatin. It has been demonstrated that this protein binds to apoptotic cells at an early stage, which raises the possibility that it is involved in dealing with apoptotic cells in vivo.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 9.5S alpha-1-glycoprotein; APCS amyloid P-component serum protein; epididymis secretory sperm binding protein Li 92n; MGC88159; Pentaxin related; pentaxin-related; Pentraxin 2; pentraxin-2; pentraxin-related; PTX 2; SAMP; SAP; Serum amyloid P-component
Gene Aliases: APCS; HEL-S-92n; PTX2; SAP
UniProt ID: (Human) P02743, (Mouse) P12246, (Rat) P23680
Entrez Gene ID: (Human) 325, (Mouse) 20219, (Rat) 29339
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support