Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Endotoxin level < 0.01EU/µg by LAL method
Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Setrusumab is under investigation in clinical trial NCT01406548 (Safety and Efficacy of Multiple Dosing Regimens of BPS804 in Post Menopausal Women With Low Bone Mineral Density).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BPS-804; MOR-05813
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support