Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product is supplied at 1 mL (100 µL/mL).
Application Notes: Functional assay - For inhibition of biological activity in vitro dilutions have to be made according to the amounts of toxin to be inactivated. ELISA - antibody can be used as detector.
Shiga-like toxins (SLTs), are also called Verotoxins. Enterohemorrhagic Escherichia coli (EHEC) strains which are primarily of serotypes 0157:H7, 026:H11, and O111:H8 have been incriminated as etiologic agents of hemorrhagic colitis and Hemolytic-uremic syndrome, a generalized disease characterized by acute renal failure, thrombocytopenia, and microangiopathic hemolytic anemia. There are several distinct E.coli SLTs. SLT-I and SLT-II are produced by EHEC. SLT-I and Shiga toxin share 99% deduced amino acid sequence homology, whereas SLT-I and SLT-II share about 60% deduced amino acid sequence homology. SLT-I and SLT-II are antigenically distinct. Antibodies to SLT-II can also neutralize a variant of SLT-II (designated SLT-Iiv) produced by strains of E.coli that cause edema. SLT-IIv is cytotoxic for Vero but not HeLa cells, distinguishing it from SLT-II. The protein structure of the toxin consists of two domains: the A polypeptide that inhibits protein synthesis by targeting ribosomes, and the B polypeptide pentamer that binds to the eukaryotic cell receptor globotriaosylceramide (Gb3) leading to receptor-mediated endocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Shiga toxin; Shigatoxin Type 2; Shigatoxin Type II; STX-2a; STX-IIa; STX2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support